Human Cluster of Differentiation 59 (CD59) ELISA Kit for detecting CD59 in serum, plasma, cell lysates, other biological fluids. Spe cies Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. 1 * 96 Tests
Catalog # MSPPEKN48000
Supplier: MARKET SOURCE PART PROCESS