Human Hepatitis A Virus Cellular Receptor 2 (HAVCR2) ELISA Kit for detecting HAVCR2 in tissue homogenates, cell lysates, cell cultur e supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sens 1 * 96 Tests
Catalog # MSPPEKN45858
Supplier: MARKET SOURCE PART PROCESS