Human Inducible T-Cell Co Stimulator (ICOS) ELISA Kit for detecting ICOS in serum, plasma, urine, cell lysates, other biological flu ids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-We 1 * 96 Tests
Catalog # MSPPEKN46031
Supplier: MARKET SOURCE PART PROCESS