Rat Epidermal Growth Factor (EGF) ELISA Kit for detecting EGF in serum, plasma, tissue homogenates, cell lysates, cell culture super nates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensit 1 * 96 Tests
Catalog # MSPPEKN45046
Supplier: MARKET SOURCE PART PROCESS