Mouse Nerve Growth Factor (NGF) ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, cell culture superna tes, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity 1 * 96 Tests
Catalog # MSPPEKN47286
Supplier: MARKET SOURCE PART PROCESS