Rat Leptin (LEP) ELISA Kit for detecting LEP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. 1 * 96 Tests
Catalog # MSPPEKN46707
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together