Human Fibroblast Growth Factor 21 (FGF21) ELISA Kit for detecting FGF21 in serum, plasma, tissue homogenates, cell lysates, cell cul ture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. 1 * 96 Tests
Catalog # MSPPEKN45268
Supplier: MARKET SOURCE PART PROCESS