Human Kidney Injury Molecule 1 (Kim1) ELISA Kit for detecting Kim1 in tissue homogenates, urine, cell culture supernates, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format 1 * 96 Tests
Catalog # MSPPEKN46600
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together