Human Tumor Necrosis Factor Alpha (TNFa) ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell cultu re supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. S 1 * 96 Tests
Catalog # MSPPEKN49057
Supplier: MARKET SOURCE PART PROCESS