ContactUsLinkComponent
Muffle Furnace L5/14/C550 Tmax1400 with flap door and protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3322
Supplier:  NABERTHERM
New Product
Muffle Furnace L5/14/C550 Tmax1400 with flap door and protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3322
Supplier:  NABERTHERM