ContactUsLinkComponent
Muffle Furnace LT5/13/C550 Tmax1300 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3205
Supplier:  NABERTHERM
New Product
Muffle Furnace LT5/13/C550 Tmax1300 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3205
Supplier:  NABERTHERM