ContactUsLinkComponent
Muffle Furnace L9/11/B510 Tmax1100 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2721
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/11/B510 Tmax1100 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2721
Supplier:  NABERTHERM