ContactUsLinkComponent
Guard-101 Gas Detector, 4 Gas Detectors for H2S, CO, LEL and O2, with Vibration, Light and Sound Alarm, 14 Hours Battery Life, Explo sion-Proof, for Work and Home, Green 1 * 1 ST
Catalog # MSPPB0DP9DBLY7
Guard-101 Gas Detector, 4 Gas Detectors for H2S, CO, LEL and O2, with Vibration, Light and Sound Alarm, 14 Hours Battery Life, Explo sion-Proof, for Work and Home, Green 1 * 1 ST
Catalog # MSPPB0DP9DBLY7