ContactUsLinkComponent
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ 1 * 1 L
Catalog # OXOIJ/7660/15
New Product
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ 1 * 1 L
Catalog # OXOIJ/7660/15