ContactUsLinkComponent
Human S100 alpha 6/PRA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human S100 alpha 6/PRA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA2830-48
Supplier:  Antibodies.com
New Product
Human S100 alpha 6/PRA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human S100 alpha 6/PRA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA2830-48
Supplier:  Antibodies.com