ContactUsLinkComponent
Human Creatine Kinase MM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Creatine Kinase MM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 96 Tests
Catalog # ANTIA2382-96
Supplier:  Antibodies.com
New Product
Human Creatine Kinase MM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Creatine Kinase MM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 96 Tests
Catalog # ANTIA2382-96
Supplier:  Antibodies.com