ContactUsLinkComponent
Rat PPAR gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat PPAR gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA1989-48
Supplier:  Antibodies.com
New Product
Rat PPAR gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat PPAR gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA1989-48
Supplier:  Antibodies.com