Mouse Serum Amyloid A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse Serum Amyloid A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 96 Tests
Catalog # ANTIA1979-96
Lieferant: Antibodies.com