Mouse Complement C4a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Complement C4a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA2023-48
Supplier: Antibodies.com