ContactUsLinkComponent
Human Retinoic Acid Receptor alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Retinoic Acid Receptor alpha in serum, plasma, tissue homogenates or other biological 1 * 48 Tests
  ANTIA2474-48
New Product
Human Retinoic Acid Receptor alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Retinoic Acid Receptor alpha in serum, plasma, tissue homogenates or other biological 1 * 48 Tests
  ANTIA2474-48