Mouse PKC epsilon ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse PKC epsilon in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2014-48
Supplier: Antibodies.com