ContactUsLinkComponent
Rat Myosin Light Chain 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Myosin Light Chain 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 96 Tests
Catalog # ANTIA270981-96
Supplier:  Antibodies.com
New Product
Rat Myosin Light Chain 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Myosin Light Chain 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 96 Tests
Catalog # ANTIA270981-96
Supplier:  Antibodies.com