ContactUsLinkComponent
Rat Surfactant Protein A/PSAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Surfactant Protein A/PSAP in serum, plasma, lung lavage fluid, tissue homogenates or other b 1 * 96 Tests
Catalog # ANTIA2010-96
Supplier:  Antibodies.com
New Product
Rat Surfactant Protein A/PSAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Surfactant Protein A/PSAP in serum, plasma, lung lavage fluid, tissue homogenates or other b 1 * 96 Tests
Catalog # ANTIA2010-96
Supplier:  Antibodies.com