ContactUsLinkComponent
Human Surfactant Protein D/SP-D ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Surfactant Protein D/SP-D in serum, plasma, tissue homogenates, cell lysates, lung lavag 1 * 96 Tests
Catalog # ANTIA2161-96
Supplier:  Antibodies.com
New Product
Human Surfactant Protein D/SP-D ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Surfactant Protein D/SP-D in serum, plasma, tissue homogenates, cell lysates, lung lavag 1 * 96 Tests
Catalog # ANTIA2161-96
Supplier:  Antibodies.com