ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Proteinase Activated Receptor 3/PAR-3 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325284-100
Supplier:  Antibodies.com
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Proteinase Activated Receptor 3/PAR-3 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325284-100
Supplier:  Antibodies.com