ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Muscarinic Acetylcholine Receptor M3/CHRM3 protein to be purified and analysed in vitro. 1 * 50 µG
Artikelnr. ANTIA325243-50
Supplier:  Antibodies.com
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Muscarinic Acetylcholine Receptor M3/CHRM3 protein to be purified and analysed in vitro. 1 * 50 µG
Artikelnr. ANTIA325243-50
Supplier:  Antibodies.com