Rat Interferon beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat Interferon beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 48 Tests
Catalog # ANTIA3112-48
Supplier: Antibodies.com