Human Carbohydrate Antigen 19-9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Carbohydrate Antigen 19-9 in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 48 Tests
Catalog # ANTIA3090-48
Supplier: Antibodies.com