Rat Interferon gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Interferon gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA2935-48
Supplier: Antibodies.com