ContactUsLinkComponent
Mouse Interferon beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse Interferon beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 48 Tests
Catalog # ANTIA3130-48
Supplier:  Antibodies.com
New Product
Mouse Interferon beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse Interferon beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 48 Tests
Catalog # ANTIA3130-48
Supplier:  Antibodies.com