ContactUsLinkComponent
Mouse Periostin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Periostin in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of samp 1 * 96 Tests
Catalog # ANTIA323234-96
Supplier:  Antibodies.com
New Product
Mouse Periostin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Periostin in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of samp 1 * 96 Tests
Catalog # ANTIA323234-96
Supplier:  Antibodies.com