ContactUsLinkComponent
Sheep IP-10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of sheep IP-10 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326512-96
Supplier:  Antibodies.com
New Product
Sheep IP-10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of sheep IP-10 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326512-96
Supplier:  Antibodies.com