ContactUsLinkComponent
Mouse Phospholipase A2 XII / PLA2G12A ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Phospholipase A2 XII / PLA2G12A in serum, plasma, and other biological fl 1 * 96 Tests
Catalog # ANTIA310456-96
Supplier:  Antibodies.com
New Product
Mouse Phospholipase A2 XII / PLA2G12A ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Phospholipase A2 XII / PLA2G12A in serum, plasma, and other biological fl 1 * 96 Tests
Catalog # ANTIA310456-96
Supplier:  Antibodies.com