Mouse P-Selectin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse P-Selectin in serum, plasma, and other biological fluids. This ELISA kit only requires 25µ l of sa 1 * 96 Tests
Catalog # ANTIA323215-96
Supplier: Antibodies.com