Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human alpha 2a Adrenergic Receptor protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325090-50
Supplier: Antibodies.com