ContactUsLinkComponent
Human Vitamin D Binding Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Vitamin D Binding Protein in serum, plasma, cell culture supernatant, cell or tissue lys 1 * 96 Tests
Catalog # ANTIA326524-96
Supplier:  Antibodies.com
New Product
Human Vitamin D Binding Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Vitamin D Binding Protein in serum, plasma, cell culture supernatant, cell or tissue lys 1 * 96 Tests
Catalog # ANTIA326524-96
Supplier:  Antibodies.com