ContactUsLinkComponent
Rat D Amino Acid Oxidase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat D Amino Acid Oxidase in serum, plasma, tissue homogenates, cell lysates or other biological fluid 1 * 48 Tests
  ANTIA2897-48
New Product
Rat D Amino Acid Oxidase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat D Amino Acid Oxidase in serum, plasma, tissue homogenates, cell lysates or other biological fluid 1 * 48 Tests
  ANTIA2897-48