ContactUsLinkComponent
Mouse E1 Ubiquitin Activating Enzyme 1 / UBA1 ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse E1 Ubiquitin Activating Enzyme 1 / UBA1 in serum, plasma, and oth 1 * 96 Tests
Catalog # ANTIA313898-96
Supplier:  Antibodies.com
New Product
Mouse E1 Ubiquitin Activating Enzyme 1 / UBA1 ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse E1 Ubiquitin Activating Enzyme 1 / UBA1 in serum, plasma, and oth 1 * 96 Tests
Catalog # ANTIA313898-96
Supplier:  Antibodies.com