Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Angiotensin II Type 2 Receptor protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325092-50
Supplier: Antibodies.com