Rat Hormone Sensitive Lipase/HSL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of rat Hormone Sensitive Lipase/HSL in serum, plasma, tissue homogenates or other biological flu 1 * 96 Tests
Catalog # ANTIA5338-96
Supplier: Antibodies.com