ContactUsLinkComponent
Human Creatine Kinase MB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Creatine Kinase MB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA4211-48
Supplier:  Antibodies.com
New Product
Human Creatine Kinase MB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Creatine Kinase MB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA4211-48
Supplier:  Antibodies.com