ContactUsLinkComponent
Mouse Aryl Hydrocarbon Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Aryl Hydrocarbon Receptor in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 96 Tests
Catalog # ANTIA4565-96
Supplier:  Antibodies.com
New Product
Mouse Aryl Hydrocarbon Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Aryl Hydrocarbon Receptor in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 96 Tests
Catalog # ANTIA4565-96
Supplier:  Antibodies.com