ContactUsLinkComponent
Mouse Insulin Degrading Enzyme/IDE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of mouse Insulin Degrading Enzyme/IDE in serum, plasma, tissue homogenates or other biological 1 * 96 Tests
  ANTIA3663-96
New Product
Mouse Insulin Degrading Enzyme/IDE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of mouse Insulin Degrading Enzyme/IDE in serum, plasma, tissue homogenates or other biological 1 * 96 Tests
  ANTIA3663-96