Rat IL-10RB/IL-10R2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat IL-10RB/IL-10R2 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5854-96
Supplier: Antibodies.com