ContactUsLinkComponent
Rabbit Superoxide Dismutase 3/EC-SOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of rabbit Superoxide Dismutase 3/EC-SOD in serum, plasma, tissue homogenates, cell lysates, 1 * 96 Tests
Catalog # ANTIA4017-96
Supplier:  Antibodies.com
New Product
Rabbit Superoxide Dismutase 3/EC-SOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of rabbit Superoxide Dismutase 3/EC-SOD in serum, plasma, tissue homogenates, cell lysates, 1 * 96 Tests
Catalog # ANTIA4017-96
Supplier:  Antibodies.com