ContactUsLinkComponent
Porcine Connexin 43/GJA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of porcine Connexin 43/GJA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
  ANTIA4386-48
New Product
Porcine Connexin 43/GJA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of porcine Connexin 43/GJA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
  ANTIA4386-48