ContactUsLinkComponent
Rat Mast Cell Chymase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of rat Mast Cell Chymase in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5887-96
Lieferant:  Antibodies.com
New Product
Rat Mast Cell Chymase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of rat Mast Cell Chymase in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5887-96
Lieferant:  Antibodies.com