Porcine S100A12/CGRP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of porcine S100A12/CGRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA82346-96
Supplier: Antibodies.com