ContactUsLinkComponent
Human Estrogen Inducible Protein pS2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Estrogen Inducible Protein pS2 in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA78859-96
Supplier:  Antibodies.com
New Product
Human Estrogen Inducible Protein pS2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Estrogen Inducible Protein pS2 in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA78859-96
Supplier:  Antibodies.com