ContactUsLinkComponent
Human ATP Binding Cassette Transporter C9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human ATP Binding Cassette Transporter C9 in tissue homogenates, cell lysates or oth 1 * 96 Tests
Catalog # ANTIA7942-96
Supplier:  Antibodies.com
New Product
Human ATP Binding Cassette Transporter C9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human ATP Binding Cassette Transporter C9 in tissue homogenates, cell lysates or oth 1 * 96 Tests
Catalog # ANTIA7942-96
Supplier:  Antibodies.com